Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C11orf54
- Entrez Gene ID
-
28970
- Gene Name
- chromosome 11 open reading frame 54
- Gene Aliases
-
PTD012, PTOD012
- Chromosome Location
-
Chr.11: 93741594 - 93765323 on Build GRCh38
View Full Product Details