Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.5: 178237616 - 178590555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173465.3 NP_775736.2 13 - 14 1131 114
XM_006714933.3 XP_006714996.1 13 - 14 861 114
XM_006714934.3 XP_006714997.1 12 - 13 834 114
XM_006714935.3 XP_006714998.1 12 - 13 825 114
XM_011534688.2 XP_011532990.1 12 - 13 834 114
XM_011534689.2 XP_011532991.1 13 - 14 861 114
XM_011534690.2 XP_011532992.1 11 - 12 807 114
XM_011534691.2 XP_011532993.1 9 - 10 753 114
XM_011534692.2 XP_011532994.1 13 - 14 1886 114
GenBank mRNA AY158895.1 - 13 - 14 1131 114
AY898961.1 - 13 - 14 1131 114
BC042428.1 - 13 - 14 810 114

View Full Product Details