Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.5: 178237616 - 178590555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173465.3 NP_775736.2 20 - 21 1566 116
XM_006714933.3 XP_006714996.1 20 - 21 1296 116
XM_006714934.3 XP_006714997.1 19 - 20 1269 116
XM_006714935.3 XP_006714998.1 19 - 20 1260 116
XM_006714936.3 XP_006714999.1 19 - 20 1233 116
XM_011534688.2 XP_011532990.1 19 - 20 1269 116
XM_011534689.2 XP_011532991.1 19 - 20 1242 116
XM_011534690.2 XP_011532992.1 18 - 19 1242 116
XM_011534691.2 XP_011532993.1 16 - 17 1188 116
XM_011534692.2 XP_011532994.1 20 - 21 2321 116
GenBank mRNA AL137461.1 - 3 - 4 193 116
AY158895.1 - 20 - 21 1566 116
AY898961.1 - 20 - 21 1566 116
BC042428.1 - 21 - 22 1348 116

View Full Product Details