The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024306.4 | NP_077282.3 | 3 - 4 | 583 | 104 |
XM_011523317.2 | XP_011521619.1 | 3 - 4 | 575 | 104 | |
XM_011523319.2 | XP_011521621.1 | 3 - 4 | 282 | 104 | |
GenBank mRNA | AJ278219.1 | - | 2 - 3 | 231 | 104 |
AK315512.1 | - | 3 - 4 | 396 | 104 | |
BC002679.2 | - | 3 - 4 | 549 | 104 | |
BC004263.1 | - | 3 - 4 | 549 | 104 | |
BC017049.1 | - | 3 - 4 | 554 | 104 |