Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRP1
Entrez Gene ID
54845
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
RBM35A, RMB35A
Chromosome Location
Chr.8: 94640999 - 94707466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034915.2 NP_001030087.2 6 - 7 830 91
NM_001122825.1 NP_001116297.1 6 - 7 830 91
NM_001122826.1 NP_001116298.1 6 - 7 830 91
NM_001122827.1 NP_001116299.1 6 - 7 830 91
NM_017697.3 NP_060167.2 6 - 7 830 91
XM_005250991.3 XP_005251048.1 6 - 7 967 91
XM_005250992.3 XP_005251049.1 6 - 7 967 91
GenBank mRNA AK000178.1 - 3 - 4 224 91
BC019932.2 - 6 - 7 791 91
BC035406.1 - 3 - 4 243 91
BC067098.1 - 6 - 7 784 91
BC099916.1 - 6 - 7 794 91
BC101621.1 - 3 - 4 189 91
BC112043.1 - 3 - 4 189 91
BX647570.1 - 6 - 7 892 91

View Full Product Details