Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESRP1
Entrez Gene ID
54845
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
RBM35A, RMB35A
Chromosome Location
Chr.8: 94640999 - 94707466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034915.2 NP_001030087.2 7 - 8 933 100
NM_001122825.1 NP_001116297.1 7 - 8 933 100
NM_001122826.1 NP_001116298.1 7 - 8 933 100
NM_001122827.1 NP_001116299.1 7 - 8 933 100
NM_017697.3 NP_060167.2 7 - 8 933 100
XM_005250991.3 XP_005251048.1 7 - 8 1070 100
XM_005250992.3 XP_005251049.1 7 - 8 1070 100
GenBank mRNA AK000178.1 - 4 - 5 327 100
BC019932.2 - 7 - 8 894 100
BC035406.1 - 4 - 5 346 100
BC067098.1 - 7 - 8 887 100
BC099916.1 - 7 - 8 897 100
BC101621.1 - 4 - 5 292 100
BC112043.1 - 4 - 5 292 100
BX647570.1 - 7 - 8 995 100

View Full Product Details