Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST5
Entrez Gene ID
6764
Gene Name
suppression of tumorigenicity 5
Gene Aliases
DENND2B, HTS1, p126
Chromosome Location
Chr.11: 8693352 - 8910951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005418.3 NP_005409.3 9 - 10 2229 117
NM_139157.2 NP_631896.1 5 - 6 697 117
NM_213618.1 NP_998783.1 6 - 7 2029 117
XM_005253077.1 XP_005253134.1 6 - 7 1944 117
XM_005253083.2 XP_005253140.1 5 - 6 839 117
XM_011520309.1 XP_011518611.1 6 - 7 1944 117
XM_011520310.1 XP_011518612.1 6 - 7 2356 117
XM_011520311.2 XP_011518613.1 6 - 7 2338 117
XM_011520312.1 XP_011518614.1 10 - 11 2303 117
XM_011520313.1 XP_011518615.1 9 - 10 2189 117
XM_011520314.1 XP_011518616.1 8 - 9 2081 117
XM_011520315.1 XP_011518617.1 8 - 9 2177 117
XM_011520316.1 XP_011518618.1 7 - 8 2107 117
XM_011520317.1 XP_011518619.1 6 - 7 2048 117
XM_011520318.1 XP_011518620.1 10 - 11 2303 117
XM_011520319.2 XP_011518621.1 6 - 7 1896 117
XM_011520320.1 XP_011518622.1 6 - 7 1944 117
XM_011520321.2 XP_011518623.1 6 - 7 1944 117
XM_011520322.1 XP_011518624.1 6 - 7 1944 117
XM_011520323.2 XP_011518625.2 5 - 6 767 117
XM_011520324.2 XP_011518626.2 5 - 6 767 117
XM_011520325.1 XP_011518627.1 5 - 6 839 117
XM_011520326.1 XP_011518628.1 9 - 10 1043 117
XM_011520327.1 XP_011518629.1 5 - 6 732 117
XM_011520328.1 XP_011518630.1 5 - 6 764 117
XM_011520329.1 XP_011518631.1 9 - 10 1043 117
XM_017018182.1 XP_016873671.1 7 - 8 2062 117
XM_017018183.1 XP_016873672.1 7 - 8 2039 117
XM_017018184.1 XP_016873673.1 9 - 10 2198 117
XM_017018185.1 XP_016873674.1 5 - 6 1994 117
XM_017018186.1 XP_016873675.1 5 - 6 1994 117
XM_017018187.1 XP_016873676.1 5 - 6 703 117
GenBank mRNA AK127763.1 - 3 - 4 1084 117
AK293245.1 - 3 - 4 462 117
AK302824.1 - 5 - 6 676 117
AK312305.1 - 6 - 7 1995 117
AK312758.1 - 5 - 6 1039 117
BC036655.1 - 9 - 10 2234 117
BX647431.1 - 6 - 7 2029 117
S45936.1 - 2 - 3 367 117
U15131.1 - 6 - 7 1957 117
U15779.1 - 2 - 3 326 117
U15780.1 - 5 - 6 697 117

View Full Product Details