Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MB21D1
Entrez Gene ID
115004
Gene Name
Mab-21 domain containing 1
Gene Aliases
C6orf150, cGAS, h-cGAS
Chromosome Location
Chr.6: 73423711 - 73452330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138441.2 NP_612450.2 2 - 3 1023 104
XM_017010232.1 XP_016865721.1 2 - 3 1033 104
GenBank mRNA AK097148.1 - 2 - 3 979 104
BC012928.1 - 2 - 3 962 104
BC108714.1 - 2 - 3 982 104
BC113606.1 - 2 - 3 959 104
BC113608.1 - 2 - 3 959 104
BC143694.1 - 2 - 3 959 104
KC294566.1 - 2 - 3 884 104

View Full Product Details