The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001693.3 | NP_001684.2 | 8 - 9 | 1022 | 77 |
GenBank mRNA | AB208806.1 | - | 8 - 9 | 812 | 77 |
AK294163.1 | - | 9 - 10 | 782 | 77 | |
AK298819.1 | - | 6 - 7 | 471 | 77 | |
AK312372.1 | - | 8 - 9 | 829 | 77 | |
BC003100.1 | - | 8 - 9 | 824 | 77 | |
BC007309.1 | - | 8 - 9 | 779 | 77 | |
BC030640.2 | - | 8 - 9 | 834 | 77 | |
L35249.1 | - | 8 - 9 | 811 | 77 | |
M60346.1 | - | 5 - 6 | 493 | 77 | |
X62949.1 | - | 8 - 9 | 833 | 77 |