The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024734.3 | NP_079010.2 | 11 - 12 | 2885 | 77 |
XM_011537158.1 | XP_011535460.1 | 11 - 12 | 2881 | 77 | |
XM_017021646.1 | XP_016877135.1 | 11 - 12 | 3051 | 77 | |
GenBank mRNA | AB047979.1 | - | 11 - 12 | 2842 | 77 |
AK023828.1 | - | 3 - 4 | 1851 | 77 | |
AK314315.1 | - | 11 - 12 | 2883 | 77 | |
BC014554.2 | - | 3 - 4 | 1425 | 77 | |
BC025251.1 | - | 3 - 4 | 1824 | 77 | |
BC068482.1 | - | 11 - 12 | 2834 | 77 |