The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003300.3 | NP_003291.2 | 7 - 8 | 946 | 69 |
NM_145725.2 | NP_663777.1 | 8 - 9 | 1085 | 69 | |
XM_017021617.1 | XP_016877106.1 | 8 - 9 | 1505 | 69 | |
XM_017021618.1 | XP_016877107.1 | 8 - 9 | 3001 | 69 | |
XM_017021620.1 | XP_016877109.1 | 8 - 9 | 1063 | 69 | |
GenBank mRNA | BC075086.2 | - | 6 - 7 | 757 | 69 |
BC075087.2 | - | 6 - 7 | 757 | 69 | |
BX247977.1 | - | 8 - 9 | 931 | 69 | |
U15637.1 | - | 8 - 9 | 939 | 69 | |
U19260.1 | - | 7 - 8 | 882 | 69 | |
U21092.1 | - | 7 - 8 | 949 | 69 |