The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171020.1 | NP_001164491.1 | 1 - 2 | 120 | 93 |
NM_152423.4 | NP_689636.3 | 1 - 2 | 120 | 93 | |
XM_011530856.1 | XP_011529158.1 | 1 - 2 | 119 | 93 | |
XM_017029269.1 | XP_016884758.1 | 1 - 2 | 119 | 93 | |
GenBank mRNA | BC031229.1 | - | 1 - 2 | 120 | 93 |
DA720504.1 | - | 1 - 2 | 109 | 93 |