Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A1
Entrez Gene ID
23446
Gene Name
solute carrier family 44 member 1
Gene Aliases
CD92, CDW92, CHTL1, CTL1
Chromosome Location
Chr.9: 105244608 - 105438504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286730.1 NP_001273659.1 14 - 15 2120 80
NM_080546.4 NP_536856.2 14 - 15 2120 80
XM_005251855.4 XP_005251912.1 14 - 15 2125 80
XM_006717027.3 XP_006717090.1 14 - 15 1933 80
XM_006717028.3 XP_006717091.1 14 - 15 1933 80
XM_006717029.2 XP_006717092.1 14 - 15 1933 80
XM_017014559.1 XP_016870048.1 14 - 15 2108 80
XM_017014560.1 XP_016870049.1 15 - 16 2098 80
GenBank mRNA AJ272365.1 - 14 - 15 2073 80
AJ420812.1 - 14 - 15 1906 80
AK022549.1 - 10 - 11 1473 80
AK124926.1 - 9 - 10 2100 80
BC018213.1 - 3 - 4 435 80
BC049203.1 - 14 - 15 1873 80

View Full Product Details