The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286730.1 | NP_001273659.1 | 2 - 3 | 377 | 70 |
NM_080546.4 | NP_536856.2 | 2 - 3 | 377 | 70 | |
XM_005251855.4 | XP_005251912.1 | 2 - 3 | 382 | 70 | |
XM_006717027.3 | XP_006717090.1 | 2 - 3 | 190 | 70 | |
XM_006717028.3 | XP_006717091.1 | 2 - 3 | 190 | 70 | |
XM_006717029.2 | XP_006717092.1 | 2 - 3 | 190 | 70 | |
XM_017014559.1 | XP_016870048.1 | 2 - 3 | 365 | 70 | |
GenBank mRNA | AJ272365.1 | - | 2 - 3 | 330 | 70 |
AJ420812.1 | - | 2 - 3 | 163 | 70 | |
BC049203.1 | - | 2 - 3 | 130 | 70 | |
CD101684.1 | - | 2 - 3 | 369 | 70 | |
CF994263.1 | - | 2 - 3 | 392 | 70 |