The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185177.1 | NP_001172106.1 | 8 - 9 | 1144 | 101 |
NM_024922.5 | NP_079198.2 | 8 - 9 | 1144 | 101 | |
XM_011522975.2 | XP_011521277.1 | 8 - 9 | 1216 | 101 | |
GenBank mRNA | AK000105.1 | - | 8 - 9 | 1083 | 101 |
AK097538.1 | - | 7 - 8 | 1586 | 101 | |
AY358609.1 | - | 8 - 9 | 1070 | 101 | |
BC053670.1 | - | 8 - 9 | 1128 | 101 |