Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 10 - 11 845 63
NM_001145658.1 NP_001139130.1 9 - 10 705 63
NM_002885.2 NP_002876.2 9 - 10 738 63
XM_005245955.3 XP_005246012.2 9 - 10 750 63
XM_006710804.2 XP_006710867.2 9 - 10 750 63
XM_006710805.2 XP_006710868.2 9 - 10 750 63
XM_017001965.1 XP_016857454.1 8 - 9 752 63
XM_017001966.1 XP_016857455.1 8 - 9 752 63
XM_017001967.1 XP_016857456.1 9 - 10 750 63
XM_017001968.1 XP_016857457.1 9 - 10 785 63
XM_017001969.1 XP_016857458.1 8 - 9 752 63
XM_017001970.1 XP_016857459.1 8 - 9 752 63
XM_017001971.1 XP_016857460.1 9 - 10 704 63
XM_017001972.1 XP_016857461.1 9 - 10 750 63
XM_017001973.1 XP_016857462.1 8 - 9 783 63
XM_017001974.1 XP_016857463.1 9 - 10 750 63
XM_017001975.1 XP_016857464.1 8 - 9 752 63
XM_017001976.1 XP_016857465.1 9 - 10 750 63
XM_017001977.1 XP_016857466.1 9 - 10 750 63
XM_017001978.1 XP_016857467.1 10 - 11 899 63
XM_017001979.1 XP_016857468.1 11 - 12 903 63
XM_017001980.1 XP_016857469.1 9 - 10 704 63
XM_017001981.1 XP_016857470.1 11 - 12 879 63
XM_017001982.1 XP_016857471.1 10 - 11 899 63
XM_017001983.1 XP_016857472.1 12 - 13 1111 63
XM_017001984.1 XP_016857473.1 11 - 12 879 63
XM_017001985.1 XP_016857474.1 9 - 10 704 63
XM_017001986.1 XP_016857475.1 11 - 12 916 63
XM_017001987.1 XP_016857476.1 10 - 11 845 63
XM_017001988.1 XP_016857477.1 10 - 11 816 63
XM_017001989.1 XP_016857478.1 11 - 12 916 63
XM_017001990.1 XP_016857479.1 10 - 11 801 63
XM_017001991.1 XP_016857480.1 10 - 11 839 63
GenBank mRNA AB007943.3 - 10 - 11 783 63
BC035030.1 - 6 - 7 494 63
BC054490.1 - 9 - 10 645 63
BM739021.1 - 2 - 3 129 63
M64788.1 - 9 - 10 681 63

View Full Product Details