Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 12 - 13 978 91
NM_001145658.1 NP_001139130.1 11 - 12 838 91
NM_002885.2 NP_002876.2 11 - 12 871 91
XM_005245955.3 XP_005246012.2 11 - 12 883 91
XM_006710804.2 XP_006710867.2 11 - 12 883 91
XM_006710805.2 XP_006710868.2 11 - 12 883 91
XM_017001965.1 XP_016857454.1 10 - 11 885 91
XM_017001966.1 XP_016857455.1 10 - 11 885 91
XM_017001967.1 XP_016857456.1 11 - 12 883 91
XM_017001968.1 XP_016857457.1 11 - 12 918 91
XM_017001969.1 XP_016857458.1 10 - 11 885 91
XM_017001970.1 XP_016857459.1 10 - 11 885 91
XM_017001971.1 XP_016857460.1 11 - 12 837 91
XM_017001972.1 XP_016857461.1 11 - 12 883 91
XM_017001973.1 XP_016857462.1 10 - 11 916 91
XM_017001974.1 XP_016857463.1 11 - 12 883 91
XM_017001975.1 XP_016857464.1 10 - 11 885 91
XM_017001976.1 XP_016857465.1 11 - 12 883 91
XM_017001977.1 XP_016857466.1 11 - 12 883 91
XM_017001978.1 XP_016857467.1 12 - 13 1032 91
XM_017001979.1 XP_016857468.1 13 - 14 1036 91
XM_017001980.1 XP_016857469.1 11 - 12 837 91
XM_017001981.1 XP_016857470.1 13 - 14 1012 91
XM_017001982.1 XP_016857471.1 12 - 13 1032 91
XM_017001983.1 XP_016857472.1 14 - 15 1244 91
XM_017001984.1 XP_016857473.1 13 - 14 1012 91
XM_017001985.1 XP_016857474.1 11 - 12 837 91
XM_017001986.1 XP_016857475.1 13 - 14 1049 91
XM_017001987.1 XP_016857476.1 12 - 13 978 91
XM_017001988.1 XP_016857477.1 12 - 13 949 91
XM_017001989.1 XP_016857478.1 13 - 14 1049 91
XM_017001990.1 XP_016857479.1 12 - 13 934 91
XM_017001991.1 XP_016857480.1 12 - 13 972 91
GenBank mRNA AB007943.3 - 12 - 13 916 91
BC035030.1 - 8 - 9 627 91
BC054490.1 - 11 - 12 778 91
BM739021.1 - 4 - 5 262 91
M64788.1 - 11 - 12 814 91

View Full Product Details