Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 16 - 17 1440 67
NM_001145658.1 NP_001139130.1 15 - 16 1300 67
NM_002885.2 NP_002876.2 15 - 16 1333 67
XM_005245955.3 XP_005246012.2 15 - 16 1345 67
XM_006710804.2 XP_006710867.2 15 - 16 1345 67
XM_006710805.2 XP_006710868.2 15 - 16 1345 67
XM_017001965.1 XP_016857454.1 14 - 15 1347 67
XM_017001966.1 XP_016857455.1 14 - 15 1347 67
XM_017001967.1 XP_016857456.1 15 - 16 1345 67
XM_017001968.1 XP_016857457.1 15 - 16 1380 67
XM_017001969.1 XP_016857458.1 14 - 15 1347 67
XM_017001970.1 XP_016857459.1 14 - 15 1347 67
XM_017001971.1 XP_016857460.1 15 - 16 1299 67
XM_017001972.1 XP_016857461.1 15 - 16 1345 67
XM_017001973.1 XP_016857462.1 14 - 15 1378 67
XM_017001974.1 XP_016857463.1 15 - 16 1345 67
XM_017001975.1 XP_016857464.1 14 - 15 1347 67
XM_017001976.1 XP_016857465.1 15 - 16 1345 67
XM_017001977.1 XP_016857466.1 15 - 16 1345 67
XM_017001978.1 XP_016857467.1 16 - 17 1494 67
XM_017001979.1 XP_016857468.1 17 - 18 1498 67
XM_017001980.1 XP_016857469.1 15 - 16 1299 67
XM_017001981.1 XP_016857470.1 17 - 18 1474 67
XM_017001982.1 XP_016857471.1 16 - 17 1494 67
XM_017001983.1 XP_016857472.1 18 - 19 1706 67
XM_017001984.1 XP_016857473.1 17 - 18 1474 67
XM_017001985.1 XP_016857474.1 15 - 16 1299 67
XM_017001986.1 XP_016857475.1 17 - 18 1511 67
XM_017001987.1 XP_016857476.1 16 - 17 1440 67
XM_017001988.1 XP_016857477.1 16 - 17 1411 67
XM_017001989.1 XP_016857478.1 17 - 18 1511 67
XM_017001990.1 XP_016857479.1 16 - 17 1396 67
XM_017001991.1 XP_016857480.1 16 - 17 1434 67
GenBank mRNA AB007943.3 - 16 - 17 1378 67
BC054490.1 - 15 - 16 1240 67
M64788.1 - 15 - 16 1276 67

View Full Product Details