Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 5 - 6 380 99
NM_001145658.1 NP_001139130.1 4 - 5 240 99
NM_002885.2 NP_002876.2 4 - 5 273 99
XM_005245955.3 XP_005246012.2 4 - 5 285 99
XM_006710804.2 XP_006710867.2 4 - 5 285 99
XM_006710805.2 XP_006710868.2 4 - 5 285 99
XM_017001965.1 XP_016857454.1 3 - 4 287 99
XM_017001966.1 XP_016857455.1 3 - 4 287 99
XM_017001967.1 XP_016857456.1 4 - 5 285 99
XM_017001968.1 XP_016857457.1 4 - 5 320 99
XM_017001969.1 XP_016857458.1 3 - 4 287 99
XM_017001970.1 XP_016857459.1 3 - 4 287 99
XM_017001971.1 XP_016857460.1 4 - 5 239 99
XM_017001972.1 XP_016857461.1 4 - 5 285 99
XM_017001973.1 XP_016857462.1 3 - 4 318 99
XM_017001974.1 XP_016857463.1 4 - 5 285 99
XM_017001975.1 XP_016857464.1 3 - 4 287 99
XM_017001976.1 XP_016857465.1 4 - 5 285 99
XM_017001977.1 XP_016857466.1 4 - 5 285 99
XM_017001978.1 XP_016857467.1 5 - 6 434 99
XM_017001979.1 XP_016857468.1 6 - 7 438 99
XM_017001980.1 XP_016857469.1 4 - 5 239 99
XM_017001981.1 XP_016857470.1 6 - 7 414 99
XM_017001982.1 XP_016857471.1 5 - 6 434 99
XM_017001983.1 XP_016857472.1 7 - 8 646 99
XM_017001984.1 XP_016857473.1 6 - 7 414 99
XM_017001985.1 XP_016857474.1 4 - 5 239 99
XM_017001986.1 XP_016857475.1 6 - 7 451 99
XM_017001987.1 XP_016857476.1 5 - 6 380 99
XM_017001988.1 XP_016857477.1 5 - 6 351 99
XM_017001989.1 XP_016857478.1 6 - 7 451 99
XM_017001990.1 XP_016857479.1 5 - 6 336 99
XM_017001991.1 XP_016857480.1 5 - 6 374 99
GenBank mRNA AB007943.3 - 5 - 6 318 99
BC054490.1 - 4 - 5 180 99
BP200029.1 - 4 - 5 240 99
BP283134.1 - 4 - 5 273 99
CV574545.1 - 4 - 5 230 99
M64788.1 - 4 - 5 216 99

View Full Product Details