Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 7 - 8 476 113
NM_001145658.1 NP_001139130.1 6 - 7 336 113
NM_002885.2 NP_002876.2 6 - 7 369 113
XM_005245955.3 XP_005246012.2 6 - 7 381 113
XM_006710804.2 XP_006710867.2 6 - 7 381 113
XM_006710805.2 XP_006710868.2 6 - 7 381 113
XM_017001965.1 XP_016857454.1 5 - 6 383 113
XM_017001966.1 XP_016857455.1 5 - 6 383 113
XM_017001967.1 XP_016857456.1 6 - 7 381 113
XM_017001968.1 XP_016857457.1 6 - 7 416 113
XM_017001969.1 XP_016857458.1 5 - 6 383 113
XM_017001970.1 XP_016857459.1 5 - 6 383 113
XM_017001971.1 XP_016857460.1 6 - 7 335 113
XM_017001972.1 XP_016857461.1 6 - 7 381 113
XM_017001973.1 XP_016857462.1 5 - 6 414 113
XM_017001974.1 XP_016857463.1 6 - 7 381 113
XM_017001975.1 XP_016857464.1 5 - 6 383 113
XM_017001976.1 XP_016857465.1 6 - 7 381 113
XM_017001977.1 XP_016857466.1 6 - 7 381 113
XM_017001978.1 XP_016857467.1 7 - 8 530 113
XM_017001979.1 XP_016857468.1 8 - 9 534 113
XM_017001980.1 XP_016857469.1 6 - 7 335 113
XM_017001981.1 XP_016857470.1 8 - 9 510 113
XM_017001982.1 XP_016857471.1 7 - 8 530 113
XM_017001983.1 XP_016857472.1 9 - 10 742 113
XM_017001984.1 XP_016857473.1 8 - 9 510 113
XM_017001985.1 XP_016857474.1 6 - 7 335 113
XM_017001986.1 XP_016857475.1 8 - 9 547 113
XM_017001987.1 XP_016857476.1 7 - 8 476 113
XM_017001988.1 XP_016857477.1 7 - 8 447 113
XM_017001989.1 XP_016857478.1 8 - 9 547 113
XM_017001990.1 XP_016857479.1 7 - 8 432 113
XM_017001991.1 XP_016857480.1 7 - 8 470 113
GenBank mRNA AB003930.1 - 4 - 5 310 113
AB007943.3 - 7 - 8 414 113
BC035030.1 - 3 - 4 125 113
BC054490.1 - 6 - 7 276 113
BP200029.1 - 6 - 7 336 113
BP283134.1 - 6 - 7 369 113
CV574545.1 - 6 - 7 326 113
M64788.1 - 6 - 7 312 113

View Full Product Details