The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006207.2 | NP_006198.1 | 6 - 7 | 119 | |
GenBank mRNA | AK289450.1 | - | 6 - 7 | 119 | |
BC010927.1 | - | 5 - 6 | 119 | ||
BQ446405.1 | - | 1 - 2 | 537 | 119 | |
CR541952.1 | - | 5 - 6 | 119 | ||
D37965.1 | - | 5 - 6 | 119 |