The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323515.1 | NP_001310444.1 | 11 - 12 | 1205 | 67 |
NM_001323516.1 | NP_001310445.1 | 11 - 12 | 1278 | 67 | |
NM_001323519.1 | NP_001310448.1 | 11 - 12 | 1327 | 67 | |
NM_001323521.1 | NP_001310450.1 | 11 - 12 | 1272 | 67 | |
NM_001323522.1 | NP_001310451.1 | 11 - 12 | 1277 | 67 | |
NM_001323523.1 | NP_001310452.1 | 11 - 12 | 1500 | 67 | |
NM_001323524.1 | NP_001310453.1 | 11 - 12 | 1278 | 67 | |
NM_001323525.1 | NP_001310454.1 | 11 - 12 | 1448 | 67 | |
NM_001323526.1 | NP_001310455.1 | 11 - 12 | 1388 | 67 | |
NM_001323528.1 | NP_001310457.1 | 11 - 12 | 1338 | 67 | |
NM_001323530.1 | NP_001310459.1 | 11 - 12 | 1388 | 67 | |
NM_001323531.1 | NP_001310460.1 | 11 - 12 | 1277 | 67 | |
NM_001323532.1 | NP_001310461.1 | 11 - 12 | 1448 | 67 | |
NM_020214.3 | NP_064599.2 | 11 - 12 | 1338 | 67 | |
NR_136594.1 | 11 - 12 | 1448 | 67 | ||
NR_136596.1 | 11 - 12 | 1591 | 67 | ||
NR_136599.1 | 11 - 12 | 1278 | 67 | ||
NR_136603.1 | 11 - 12 | 1272 | 67 | ||
NR_136604.1 | 11 - 12 | 1388 | 67 | ||
NR_136605.1 | 11 - 12 | 1388 | 67 | ||
NR_136606.1 | 11 - 12 | 1500 | 67 | ||
NR_136607.1 | 11 - 12 | 1448 | 67 | ||
NR_136608.1 | 11 - 12 | 1338 | 67 | ||
NR_136609.1 | 11 - 12 | 1448 | 67 | ||
NR_136610.1 | 11 - 12 | 1448 | 67 | ||
NR_136611.1 | 11 - 12 | 1388 | 67 | ||
XM_011521805.1 | XP_011520107.1 | 6 - 7 | 710 | 67 | |
XM_017022418.1 | XP_016877907.1 | 10 - 11 | 1928 | 67 | |
GenBank mRNA | AB499727.1 | - | 8 - 9 | 806 | 67 |
AB499728.1 | - | 8 - 9 | 806 | 67 | |
AK022547.1 | - | 10 - 11 | 1029 | 67 | |
AK126416.1 | - | 8 - 9 | 1571 | 67 | |
AL122091.1 | - | 6 - 7 | 620 | 67 | |
AL390087.1 | - | 8 - 9 | 949 | 67 | |
BC015314.1 | - | 7 - 8 | 604 | 67 | |
BC026955.1 | - | 7 - 8 | 603 | 67 | |
BC110902.1 | - | 9 - 10 | 854 | 67 | |
BC119688.2 | - | 9 - 10 | 865 | 67 | |
BC119689.1 | - | 9 - 10 | 865 | 67 |