The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323515.1 | NP_001310444.1 | 22 - 23 | 2106 | 71 |
NM_001323516.1 | NP_001310445.1 | 22 - 23 | 2179 | 71 | |
NM_001323519.1 | NP_001310448.1 | 22 - 23 | 2225 | 71 | |
NM_001323521.1 | NP_001310450.1 | 22 - 23 | 2170 | 71 | |
NM_001323522.1 | NP_001310451.1 | 22 - 23 | 2175 | 71 | |
NM_001323523.1 | NP_001310452.1 | 22 - 23 | 2398 | 71 | |
NM_001323524.1 | NP_001310453.1 | 22 - 23 | 2176 | 71 | |
NM_001323525.1 | NP_001310454.1 | 22 - 23 | 2349 | 71 | |
NM_001323526.1 | NP_001310455.1 | 22 - 23 | 2286 | 71 | |
NM_001323528.1 | NP_001310457.1 | 22 - 23 | 2239 | 71 | |
NM_001323530.1 | NP_001310459.1 | 22 - 23 | 2289 | 71 | |
NM_001323531.1 | NP_001310460.1 | 22 - 23 | 2178 | 71 | |
NM_001323532.1 | NP_001310461.1 | 22 - 23 | 2346 | 71 | |
NM_020214.3 | NP_064599.2 | 22 - 23 | 2236 | 71 | |
NR_136594.1 | 22 - 23 | 2397 | 71 | ||
NR_136596.1 | 21 - 22 | 2419 | 71 | ||
NR_136599.1 | 21 - 22 | 2109 | 71 | ||
NR_136603.1 | 21 - 22 | 2100 | 71 | ||
NR_136604.1 | 23 - 24 | 2407 | 71 | ||
NR_136605.1 | 20 - 21 | 2106 | 71 | ||
NR_136606.1 | 21 - 22 | 2328 | 71 | ||
NR_136607.1 | 21 - 22 | 2276 | 71 | ||
NR_136608.1 | 21 - 22 | 2166 | 71 | ||
NR_136609.1 | 20 - 21 | 2166 | 71 | ||
NR_136610.1 | 21 - 22 | 2279 | 71 | ||
NR_136611.1 | 21 - 22 | 2216 | 71 | ||
XM_011521805.1 | XP_011520107.1 | 17 - 18 | 1611 | 71 | |
XM_017022418.1 | XP_016877907.1 | 21 - 22 | 2826 | 71 | |
GenBank mRNA | AK022547.1 | - | 20 - 21 | 1860 | 71 |
AK091172.1 | - | 2 - 3 | 3231 | 71 | |
AK126416.1 | - | 17 - 18 | 2289 | 71 | |
AL122091.1 | - | 17 - 18 | 1518 | 71 | |
AL390087.1 | - | 20 - 21 | 1896 | 71 | |
AL390093.1 | - | 8 - 9 | 554 | 71 | |
BC015176.1 | - | 5 - 6 | 301 | 71 | |
BC015314.1 | - | 18 - 19 | 1505 | 71 | |
BC026955.1 | - | 17 - 18 | 1431 | 71 | |
BC110902.1 | - | 20 - 21 | 1752 | 71 |