Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRG1
Entrez Gene ID
9289
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location
Chr.16: 57619535 - 57665039 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145770.2 NP_001139242.1 5 - 6 911 94
NM_001145771.2 NP_001139243.1 5 - 6 1349 94
NM_001145772.2 NP_001139244.1 5 - 6 1375 94
NM_001145773.2 NP_001139245.1 4 - 5 1246 94
NM_001145774.2 NP_001139246.1 5 - 6 1148 94
NM_001290143.1 NP_001277072.1 4 - 5 800 94
NM_001290144.1 NP_001277073.1 4 - 5 987 94
NM_005682.6 NP_005673.3 5 - 6 911 94
NM_201524.3 NP_958932.1 5 - 6 1353 94
NM_201525.3 NP_958933.1 4 - 5 1231 94
XM_005256237.2 XP_005256294.1 4 - 5 947 94
XM_005256238.2 XP_005256295.1 5 - 6 1286 94
XM_005256239.2 XP_005256296.1 5 - 6 1048 94
XM_005256240.4 XP_005256297.1 4 - 5 769 94
XM_005256241.4 XP_005256298.1 4 - 5 773 94
XM_005256242.4 XP_005256299.1 4 - 5 830 94
XM_005256244.3 XP_005256301.1 4 - 5 922 94
XM_005256245.2 XP_005256302.1 4 - 5 973 94
XM_005256246.2 XP_005256303.1 5 - 6 1033 94
XM_005256247.2 XP_005256304.1 4 - 5 932 94
XM_005256248.2 XP_005256305.1 4 - 5 997 94
XM_005256249.3 XP_005256306.1 4 - 5 788 94
XM_005256251.4 XP_005256308.1 4 - 5 814 94
XM_005256252.2 XP_005256309.1 4 - 5 758 94
XM_005256254.2 XP_005256311.1 4 - 5 958 94
XM_005256255.2 XP_005256312.1 5 - 6 1271 94
XM_006721338.2 XP_006721401.1 5 - 6 1069 94
XM_006721339.3 XP_006721402.1 5 - 6 876 94
XM_006721340.3 XP_006721403.1 5 - 6 1134 94
XM_006721341.2 XP_006721404.1 5 - 6 1091 94
XM_006721342.2 XP_006721405.1 5 - 6 1054 94
XM_006721343.3 XP_006721406.1 5 - 6 1119 94
XM_006721344.2 XP_006721407.1 5 - 6 1076 94
XM_006721345.3 XP_006721408.1 5 - 6 845 94
XM_006721346.2 XP_006721409.1 4 - 5 700 94
XM_006721347.2 XP_006721410.1 5 - 6 1286 94
XM_011523461.2 XP_011521763.1 5 - 6 1037 94
XM_011523462.2 XP_011521764.1 4 - 5 852 94
XM_011523463.2 XP_011521765.1 5 - 6 862 94
XM_011523464.2 XP_011521766.1 5 - 6 1022 94
XM_011523465.2 XP_011521767.1 5 - 6 960 94
XM_011523466.2 XP_011521768.1 4 - 5 1007 94
XM_011523467.2 XP_011521769.1 5 - 6 754 94
XM_011523468.2 XP_011521770.1 4 - 5 756 94
XM_017023892.1 XP_016879381.1 4 - 5 958 94
GenBank mRNA AF106858.1 - 4 - 5 781 94
AJ011001.1 - 5 - 6 935 94
AK075270.1 - 5 - 6 863 94
AK075555.1 - 5 - 6 886 94
AK297236.1 - 4 - 5 860 94
AK299110.1 - 4 - 5 775 94
AK312405.1 - 4 - 5 1067 94
AY358400.1 - 5 - 6 832 94
BC008770.2 - 4 - 5 777 94
BC013207.1 - 5 - 6 925 94
BT007311.1 - 3 - 4 619 94
CR936747.1 - 4 - 5 742 94
EU432119.1 - 3 - 4 619 94
KU178472.1 - 3 - 4 619 94
KU178473.1 - 3 - 4 619 94

View Full Product Details