Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRG1
Entrez Gene ID
9289
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location
Chr.16: 57619535 - 57665039 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145770.2 NP_001139242.1 6 - 7 1064 65
NM_001145771.2 NP_001139243.1 6 - 7 1502 65
NM_001145772.2 NP_001139244.1 6 - 7 1528 65
NM_001145773.2 NP_001139245.1 5 - 6 1399 65
NM_001145774.2 NP_001139246.1 6 - 7 1301 65
NM_001290142.1 NP_001277071.1 5 - 6 604 65
NM_001290143.1 NP_001277072.1 5 - 6 953 65
NM_001290144.1 NP_001277073.1 5 - 6 1140 65
NM_005682.6 NP_005673.3 6 - 7 1064 65
NM_201524.3 NP_958932.1 6 - 7 1506 65
NM_201525.3 NP_958933.1 5 - 6 1384 65
XM_005256237.2 XP_005256294.1 5 - 6 1100 65
XM_005256238.2 XP_005256295.1 6 - 7 1439 65
XM_005256239.2 XP_005256296.1 6 - 7 1201 65
XM_005256240.4 XP_005256297.1 5 - 6 922 65
XM_005256241.4 XP_005256298.1 5 - 6 926 65
XM_005256242.4 XP_005256299.1 5 - 6 983 65
XM_005256244.3 XP_005256301.1 5 - 6 1075 65
XM_005256245.2 XP_005256302.1 5 - 6 1126 65
XM_005256246.2 XP_005256303.1 6 - 7 1186 65
XM_005256247.2 XP_005256304.1 5 - 6 1085 65
XM_005256248.2 XP_005256305.1 5 - 6 1150 65
XM_005256249.3 XP_005256306.1 5 - 6 941 65
XM_005256251.4 XP_005256308.1 5 - 6 967 65
XM_005256252.2 XP_005256309.1 5 - 6 911 65
XM_005256254.2 XP_005256311.1 5 - 6 1111 65
XM_005256255.2 XP_005256312.1 6 - 7 1424 65
XM_006721338.2 XP_006721401.1 6 - 7 1222 65
XM_006721339.3 XP_006721402.1 6 - 7 1029 65
XM_006721340.3 XP_006721403.1 6 - 7 1287 65
XM_006721341.2 XP_006721404.1 6 - 7 1244 65
XM_006721342.2 XP_006721405.1 6 - 7 1207 65
XM_006721343.3 XP_006721406.1 6 - 7 1272 65
XM_006721344.2 XP_006721407.1 6 - 7 1229 65
XM_006721345.3 XP_006721408.1 6 - 7 998 65
XM_006721346.2 XP_006721409.1 5 - 6 853 65
XM_006721347.2 XP_006721410.1 6 - 7 1439 65
XM_011523461.2 XP_011521763.1 6 - 7 1190 65
XM_011523462.2 XP_011521764.1 5 - 6 1005 65
XM_011523463.2 XP_011521765.1 6 - 7 1015 65
XM_011523464.2 XP_011521766.1 6 - 7 1175 65
XM_011523465.2 XP_011521767.1 6 - 7 1113 65
XM_011523466.2 XP_011521768.1 5 - 6 1160 65
XM_011523467.2 XP_011521769.1 6 - 7 907 65
XM_011523468.2 XP_011521770.1 5 - 6 909 65
XM_017023892.1 XP_016879381.1 5 - 6 1111 65
GenBank mRNA AF106858.1 - 5 - 6 934 65
AJ011001.1 - 6 - 7 1088 65
AK075270.1 - 6 - 7 1016 65
AK075555.1 - 6 - 7 1039 65
AK131550.1 - 5 - 6 559 65
AK297236.1 - 5 - 6 1013 65
AK299110.1 - 5 - 6 928 65
AK312405.1 - 5 - 6 1220 65
AY358400.1 - 6 - 7 985 65
BC008770.2 - 5 - 6 930 65
BC013207.1 - 6 - 7 1078 65
BT007311.1 - 4 - 5 772 65
CR936747.1 - 5 - 6 895 65
EU432119.1 - 4 - 5 772 65
KU178472.1 - 4 - 5 772 65
KU178473.1 - 4 - 5 772 65

View Full Product Details