The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014708.4 | NP_055523.1 | 30 - 31 | 2883 | 111 |
XM_006719706.2 | XP_006719769.1 | 30 - 31 | 2865 | 111 | |
XM_011539027.2 | XP_011537329.1 | 30 - 31 | 2865 | 111 | |
XM_011539029.2 | XP_011537331.1 | 30 - 31 | 2865 | 111 | |
XM_011539030.1 | XP_011537332.1 | 30 - 31 | 2865 | 111 | |
XM_017020254.1 | XP_016875743.1 | 30 - 31 | 2865 | 111 | |
XM_017020255.1 | XP_016875744.1 | 29 - 30 | 2829 | 111 | |
XM_017020256.1 | XP_016875745.1 | 11 - 12 | 1139 | 111 | |
XM_017020257.1 | XP_016875746.1 | 30 - 31 | 2865 | 111 | |
GenBank mRNA | BC150278.1 | - | 30 - 31 | 2883 | 111 |
D79988.1 | - | 30 - 31 | 2883 | 111 |