The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014708.4 | NP_055523.1 | 45 - 46 | 4817 | 101 |
XM_006719706.2 | XP_006719769.1 | 45 - 46 | 4799 | 101 | |
XM_011539027.2 | XP_011537329.1 | 45 - 46 | 4799 | 101 | |
XM_017020255.1 | XP_016875744.1 | 44 - 45 | 4763 | 101 | |
XM_017020256.1 | XP_016875745.1 | 26 - 27 | 3073 | 101 | |
GenBank mRNA | AK293403.1 | - | 2 - 3 | 305 | 101 |
BC150278.1 | - | 45 - 46 | 4817 | 101 | |
D79988.1 | - | 45 - 46 | 4817 | 101 |