The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014708.4 | NP_055523.1 | 51 - 52 | 5663 | 80 |
XM_006719706.2 | XP_006719769.1 | 51 - 52 | 5645 | 80 | |
XM_011539027.2 | XP_011537329.1 | 51 - 52 | 5645 | 80 | |
XM_017020254.1 | XP_016875743.1 | 50 - 51 | 5555 | 80 | |
XM_017020255.1 | XP_016875744.1 | 50 - 51 | 5609 | 80 | |
XM_017020256.1 | XP_016875745.1 | 32 - 33 | 3919 | 80 | |
GenBank mRNA | AK093470.1 | - | 6 - 7 | 1019 | 80 |
AK293403.1 | - | 8 - 9 | 1151 | 80 | |
BC150278.1 | - | 51 - 52 | 5663 | 80 | |
D79988.1 | - | 51 - 52 | 5663 | 80 |