Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNTC1
Entrez Gene ID
9735
Gene Name
kinetochore associated 1
Gene Aliases
ROD
Chromosome Location
Chr.12: 122527114 - 122626409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014708.4 NP_055523.1 55 - 56 6036 76
XM_006719706.2 XP_006719769.1 55 - 56 6018 76
XM_011539027.2 XP_011537329.1 55 - 56 5940 76
XM_017020254.1 XP_016875743.1 54 - 55 5928 76
XM_017020255.1 XP_016875744.1 54 - 55 5982 76
XM_017020256.1 XP_016875745.1 36 - 37 4292 76
GenBank mRNA AK093470.1 - 10 - 11 1392 76
AK293403.1 - 12 - 13 1524 76
AK303337.1 - 30 - 31 2793 76
BC150278.1 - 55 - 56 6036 76
D79988.1 - 55 - 56 6036 76

View Full Product Details