Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROR1
Entrez Gene ID
4919
Gene Name
receptor tyrosine kinase like orphan receptor 1
Gene Aliases
NTRKR1, dJ537F10.1
Chromosome Location
Chr.1: 63774019 - 64181498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083592.1 NP_001077061.1 6 - 7 1332 97
NM_005012.3 NP_005003.2 6 - 7 1332 97
XM_011541526.1 XP_011539828.1 6 - 7 931 97
XM_017001376.1 XP_016856865.1 5 - 6 1232 97
XM_017001377.1 XP_016856866.1 8 - 9 3875 97
GenBank mRNA AK308088.1 - 6 - 7 1338 97
AK309189.1 - 3 - 4 574 97
BC080541.1 - 6 - 7 1332 97
BC128386.1 - 7 - 8 942 97
M97675.1 - 6 - 7 1308 97

View Full Product Details