The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006558.2 | NP_006549.1 | 6 - 7 | 1213 | 129 |
XM_005250757.2 | XP_005250814.1 | 6 - 7 | 905 | 129 | |
XM_005250758.3 | XP_005250815.1 | 6 - 7 | 1217 | 129 | |
XM_011516796.2 | XP_011515098.1 | 6 - 7 | 1055 | 129 | |
XM_011516797.2 | XP_011515099.1 | 6 - 7 | 1090 | 129 | |
GenBank mRNA | AF051321.1 | - | 6 - 7 | 1205 | 129 |
AF069681.1 | - | 6 - 7 | 1161 | 129 | |
BC032606.2 | - | 6 - 7 | 1222 | 129 | |
BC068536.1 | - | 7 - 8 | 1014 | 129 |