Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP25
Entrez Gene ID
6616
Gene Name
synaptosome associated protein 25
Gene Aliases
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
Chromosome Location
Chr.20: 10218694 - 10307420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322903.1 NP_001309832.1 6 - 7 647 122
NM_001322904.1 NP_001309833.1 6 - 7 617 122
NM_001322905.1 NP_001309834.1 6 - 7 580 122
NM_001322906.1 NP_001309835.1 6 - 7 574 122
NM_001322907.1 NP_001309836.1 7 - 8 775 122
NM_001322908.1 NP_001309837.1 7 - 8 698 122
NM_001322909.1 NP_001309838.1 6 - 7 565 122
NM_001322910.1 NP_001309839.1 6 - 7 672 122
NM_130811.3 NP_570824.1 5 - 6 520 122
XM_005260808.4 XP_005260865.1 5 - 6 598 122
XM_017028023.1 XP_016883512.1 5 - 6 624 122
GenBank mRNA AK094560.1 - 6 - 7 606 122
AK098347.1 - 6 - 7 393 122
AK223617.1 - 5 - 6 482 122
AK314359.1 - 5 - 6 431 122
BI552777.1 - 5 - 6 493 122
BI602062.1 - 5 - 6 499 122
BX647443.1 - 5 - 6 392 122
D21267.1 - 5 - 6 482 122
L19761.1 - 5 - 6 365 122

View Full Product Details