The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303037.1 | NP_001289966.1 | 4 - 5 | 671 | 154 |
NM_016447.3 | NP_057531.2 | 5 - 6 | 840 | 154 | |
XM_005249775.4 | XP_005249832.1 | 3 - 4 | 452 | 154 | |
XM_006715738.3 | XP_006715801.1 | 4 - 5 | 971 | 154 | |
XM_006715739.3 | XP_006715802.1 | 4 - 5 | 562 | 154 | |
XM_006715740.3 | XP_006715803.1 | 4 - 5 | 559 | 154 | |
XM_011515425.2 | XP_011513727.1 | 4 - 5 | 667 | 154 | |
XM_017012315.1 | XP_016867804.1 | 4 - 5 | 595 | 154 | |
XM_017012316.1 | XP_016867805.1 | 4 - 5 | 716 | 154 | |
XM_017012317.1 | XP_016867806.1 | 4 - 5 | 596 | 154 | |
GenBank mRNA | AF162130.2 | - | 4 - 5 | 612 | 154 |
AL136836.1 | - | 5 - 6 | 720 | 154 | |
BC023638.2 | - | 4 - 5 | 635 | 154 |