The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005418.3 | NP_005409.3 | 2 - 3 | 233 | 112 |
GenBank mRNA | BC036655.1 | - | 2 - 3 | 238 | 112 |