The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000970.4 | NP_000961.2 | 5 - 6 | 586 | 77 |
NM_001024662.2 | NP_001019833.1 | 5 - 6 | 762 | 77 | |
NM_001320137.1 | NP_001307066.1 | 5 - 6 | 767 | 77 | |
NM_001320138.1 | NP_001307067.1 | 5 - 6 | 801 | 77 | |
NM_001320139.1 | NP_001307068.1 | 5 - 6 | 648 | 77 | |
NM_001320140.1 | NP_001307069.1 | 5 - 6 | 810 | 77 | |
NM_001320141.1 | NP_001307070.1 | 7 - 8 | 875 | 77 | |
NM_001320142.1 | NP_001307071.1 | 5 - 6 | 522 | 77 | |
XM_017019781.1 | XP_016875270.1 | 7 - 8 | 1910 | 77 | |
XM_017019782.1 | XP_016875271.1 | 7 - 8 | 1071 | 77 | |
GenBank mRNA | AF261087.1 | - | 1 - 1 | 556 | 77 |
AK299470.1 | - | 6 - 7 | 516 | 77 | |
AK311863.1 | - | 5 - 6 | 550 | 77 | |
AY320410.1 | - | 2 - 3 | 106 | 77 | |
BC004138.2 | - | 1 - 1 | 562 | 77 | |
BC013863.1 | - | 1 - 1 | 536 | 77 | |
BC020679.1 | - | 1 - 1 | 550 | 77 | |
BC022444.1 | - | 5 - 6 | 717 | 77 | |
BC031009.1 | - | 1 - 1 | 550 | 77 | |
BC032299.1 | - | 1 - 1 | 547 | 77 | |
BC071912.1 | - | 1 - 1 | 539 | 77 | |
BC104824.1 | - | 5 - 6 | 562 | 77 | |
BC104826.1 | - | 5 - 6 | 562 | 77 | |
BG030827.1 | - | 1 - 1 | 312 | 77 | |
CD049618.1 | - | 5 - 6 | 732 | 77 | |
D17554.1 | - | 5 - 6 | 552 | 77 | |
S71022.1 | - | 4 - 5 | 76 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK126570.1 | - | - | 1298 | 77 |