Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMF1
Entrez Gene ID
9491
Gene Name
proteasome inhibitor subunit 1
Gene Aliases
PI31
Chromosome Location
Chr.20: 1113229 - 1172246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323407.1 NP_001310336.1 3 - 4 423 68
NM_001323408.1 NP_001310337.1 3 - 4 552 68
NM_001323409.1 NP_001310338.1 3 - 4 552 68
NM_001323410.1 NP_001310339.1 3 - 4 552 68
NM_006814.4 NP_006805.2 3 - 4 552 68
NM_178578.3 NP_848693.2 4 - 5 573 68
GenBank mRNA AK300667.1 - 3 - 4 297 68
AK302164.1 - 3 - 4 389 68
BC126462.1 - 4 - 5 429 68
BI830805.1 - 4 - 5 506 68
D88378.1 - 3 - 4 497 68
DB028624.1 - 4 - 5 539 68
DC398121.1 - 3 - 4 423 68

View Full Product Details