Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MTRF1
- Entrez Gene ID
-
9617
- Gene Name
- mitochondrial translational release factor 1
- Gene Aliases
-
MRF1, MTTRF1, RF1
- Chromosome Location
-
Chr.13: 41216369 - 41311794 on Build GRCh38
View Full Product Details