Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10A3
Entrez Gene ID
26496
Gene Name
olfactory receptor family 10 subfamily A member 3
Gene Aliases
HSHTPCRX12, HTPCRX12
Chromosome Location
Chr.11: 7938576 - 7939520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003745.1 NP_001003745.1 1 - 1 852 76
GenBank mRNA BC137031.1 - 1 - 1 926 76
BC137032.1 - 1 - 1 926 76

View Full Product Details