The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290104.1 | NP_001277033.1 | 3 - 4 | 637 | 114 |
NM_003460.2 | NP_003451.1 | 3 - 4 | 637 | 114 | |
XM_011545942.2 | XP_011544244.1 | 2 - 3 | 166 | 114 | |
GenBank mRNA | AK293805.1 | - | 2 - 3 | 181 | 114 |
AK308820.1 | - | 2 - 3 | 181 | 114 | |
AK310454.1 | - | 3 - 4 | 637 | 114 | |
AK313003.1 | - | 2 - 3 | 179 | 114 | |
BC096304.3 | - | 2 - 3 | 154 | 114 | |
BC096305.3 | - | 2 - 3 | 154 | 114 | |
BC096306.1 | - | 2 - 3 | 154 | 114 | |
BC096307.3 | - | 2 - 3 | 154 | 114 | |
M90366.1 | - | 2 - 3 | 154 | 114 |