Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZP2
Entrez Gene ID
7783
Gene Name
zona pellucida glycoprotein 2
Gene Aliases
ZPA, Zp-2
Chromosome Location
Chr.16: 21197452 - 21214510 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290104.1 NP_001277033.1 6 - 7 963 77
NM_003460.2 NP_003451.1 6 - 7 963 77
XM_011545942.2 XP_011544244.1 5 - 6 492 77
GenBank mRNA AK308820.1 - 5 - 6 507 77
AK310454.1 - 6 - 7 963 77
AK313003.1 - 5 - 6 505 77
BC096304.3 - 5 - 6 480 77
BC096305.3 - 5 - 6 480 77
BC096306.1 - 5 - 6 480 77
BC096307.3 - 5 - 6 480 77
M90366.1 - 5 - 6 480 77

View Full Product Details