Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
912
Gene Name
CD1d molecule
Gene Aliases
CD1A, R3, R3G1
Chromosome Location
Chr.1: 158178038 - 158186427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001766.3 NP_001757.1 5 - 6 1381 63
XM_005245583.1 XP_005245640.1 4 - 5 1060 63
XM_006711621.1 XP_006711684.1 4 - 5 942 63
XM_011510125.1 XP_011508427.1 5 - 6 1196 63
XM_011510127.1 XP_011508429.1 4 - 5 1024 63
GenBank mRNA BC027926.1 - 5 - 6 1381 63
J04142.1 - 4 - 5 1046 63

View Full Product Details