The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017416.1 | NP_059112.1 | 9 - 10 | 1948 | 110 |
XM_011530905.2 | XP_011529207.1 | 7 - 8 | 956 | 110 | |
XM_017029403.1 | XP_016884892.1 | 9 - 10 | 1366 | 110 | |
GenBank mRNA | AF181285.1 | - | 7 - 8 | 1110 | 110 |
AF212016.1 | - | 8 - 9 | 1192 | 110 | |
AF284436.1 | - | 9 - 10 | 1343 | 110 | |
AJ272208.1 | - | 9 - 10 | 1948 | 110 | |
AJ290436.1 | - | 8 - 9 | 1211 | 110 | |
AK290196.1 | - | 9 - 10 | 2064 | 110 | |
BC104784.1 | - | 9 - 10 | 1240 | 110 | |
BC104786.1 | - | 9 - 10 | 1240 | 110 |