The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017416.1 | NP_059112.1 | 10 - 11 | 2116 | 125 |
XM_011530905.2 | XP_011529207.1 | 8 - 9 | 1124 | 125 | |
XM_017029403.1 | XP_016884892.1 | 10 - 11 | 1534 | 125 | |
GenBank mRNA | AF181285.1 | - | 8 - 9 | 1278 | 125 |
AF212016.1 | - | 9 - 10 | 1360 | 125 | |
AF284436.1 | - | 10 - 11 | 1511 | 125 | |
AJ272208.1 | - | 10 - 11 | 2116 | 125 | |
AJ290436.1 | - | 9 - 10 | 1379 | 125 | |
AK290196.1 | - | 10 - 11 | 2232 | 125 | |
AK297237.1 | - | 2 - 3 | 412 | 125 | |
BC104784.1 | - | 10 - 11 | 1408 | 125 | |
BC104786.1 | - | 10 - 11 | 1408 | 125 |