The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290145.1 | NP_001277074.1 | 8 - 9 | 878 | 70 |
NM_015584.4 | NP_056399.1 | 8 - 9 | 932 | 70 | |
GenBank mRNA | AF077203.1 | - | 8 - 9 | 840 | 70 |
AF179891.1 | - | 8 - 9 | 791 | 70 | |
AK293707.1 | - | 8 - 9 | 752 | 70 | |
AK313232.1 | - | 8 - 9 | 824 | 70 | |
BC000655.2 | - | 8 - 9 | 824 | 70 | |
BC009265.2 | - | 8 - 9 | 818 | 70 | |
BC018864.2 | - | 8 - 9 | 857 | 70 |