Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LST1
Entrez Gene ID
7940
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144, D6S49E, LST-1
Chromosome Location
Chr.6: 31586179 - 31588909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205837.2 NP_995309.2 3 - 4 325 88
GenBank mRNA AF000424.1 - 3 - 4 325 88

View Full Product Details