The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129765.1 | NP_001123237.1 | 1 - 2 | 153 | 63 |
XM_011531178.2 | XP_011529480.1 | 1 - 2 | 119 | 63 |