The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129765.1 | NP_001123237.1 | 5 - 6 | 675 | 75 |
NM_015922.2 | NP_057006.1 | 4 - 5 | 608 | 75 | |
XM_011531178.2 | XP_011529480.1 | 6 - 7 | 735 | 75 | |
XM_017029564.1 | XP_016885053.1 | 4 - 5 | 540 | 75 | |
GenBank mRNA | AK026549.1 | - | 4 - 5 | 498 | 75 |
BC000245.1 | - | 4 - 5 | 529 | 75 | |
BC007816.2 | - | 4 - 5 | 579 | 75 | |
U47105.2 | - | 4 - 5 | 608 | 75 |