The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323640.1 | NP_001310569.1 | 3 - 4 | 1541 | 89 |
NM_024063.2 | NP_076968.2 | 3 - 4 | 1541 | 89 | |
NR_027635.1 | 3 - 4 | 1541 | 89 | ||
NR_136645.1 | 3 - 4 | 1548 | 89 | ||
NR_136646.1 | 4 - 5 | 1736 | 89 | ||
NR_136647.1 | 4 - 5 | 1703 | 89 | ||
NR_136648.1 | 4 - 5 | 1736 | 89 | ||
GenBank mRNA | AK023232.1 | - | 3 - 4 | 1487 | 89 |
AK225463.1 | - | 3 - 4 | 1487 | 89 | |
AK291457.1 | - | 3 - 4 | 1494 | 89 | |
BC000981.2 | - | 3 - 4 | 1474 | 89 | |
BC051861.2 | - | 3 - 4 | 1486 | 89 |