The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005921.1 | NP_005912.1 | 10 - 11 | 2467 | 84 |
XM_017009484.1 | XP_016864973.1 | 10 - 11 | 5178 | 84 | |
XM_017009485.1 | XP_016864974.1 | 10 - 11 | 1906 | 84 | |
GenBank mRNA | AF042838.1 | - | 10 - 11 | 1909 | 84 |
AK307828.1 | - | 11 - 12 | 1687 | 84 |