The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080435.2 | NP_001073904.1 | 8 - 9 | 1741 | 157 |
XM_005272421.4 | XP_005272478.1 | 8 - 9 | 1357 | 157 | |
XM_005272422.3 | XP_005272479.1 | 8 - 9 | 1104 | 157 | |
XM_005272423.4 | XP_005272480.1 | 8 - 9 | 1343 | 157 | |
XM_011521232.2 | XP_011519534.1 | 6 - 7 | 1553 | 157 | |
GenBank mRNA | AB075851.1 | - | 8 - 9 | 1727 | 157 |
AK126887.1 | - | 8 - 9 | 1743 | 157 |